1-Ethylpropylzinc bromide 0.5 M in THF

Danger


Packaged under Argon in resealable ChemSeal™ bottles.

H58214-AD
AAH58214-ADEA 222.56 USD
AAH58214-AD
1-Ethylpropylzinc bromide 0.5 M in THF
1-Ethylpropylzinc bromide
Formula: C₅H₁₁BrZn
Density: 0.95
Storage Temperature: Ambient
MDL Number: MFCD01311410
CAS Number: 308796-09-0
UN: 3399
ADR: "4.3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR