Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Dichlorophenylzinc iodide 0.5 M in THF

Supplier: Alfa Aesar
Danger

Packaged under Argon in resealable ChemSeal™ bottles.

H58050-AD
AAH58050-ADEA 177.16 USD InStock
AAH58050-AD
3,5-Dichlorophenylzinc iodide 0.5 M in THF
3,5-Dichlorophenylzinc iodide


Formula: C₆H₃Cl₂IZn
MW: 338.274 g/mol
Density: 1,002
Storage Temperature: Refrigerator
MDL Number: MFCD01319865
CAS Number: 312692-86-7
UN: 3399
ADR: ,
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty