4-(3-Ethyl-1,2,4-oxadiazol-5-yl)benzonitrile 97%
|
Warning
|
Synonyms:
Benzonitrile, 4-(3-ethyl-1,2,4-oxadiazol-5-yl)-
H57257-03
H57257-MD
AAH57257-03EA
147.29
USD
AAH57257-03
AAH57257-MD
4-(3-Ethyl-1,2,4-oxadiazol-5-yl)benzonitrile 97%
4-(3-Ethyl-1,2,4-oxadiazol-5-yl)benzonitrile
Formula:
C₁₁H₉N₃O MW: 199.21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22682905 CAS Number: 1421262-65-8 UN: 3439 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...