N-Boc-N-methylethylenediamine 97+%
|
Danger
|
Synonyms:
N-Boc-(2-Aminoethyl)-N-methylamine
, N-Methylethane-1,2-diamine, N-BOC protected
H56456-AA
H56456-XF
AAH56456-AAEA
950.25
USD
AAH56456-AA
AAH56456-XF
N-Boc-N-methylethylenediamine 97+%
N-Boc-N-methyl-ethylenediamine
Formula:
C₈H₁₈N₂O₂ MW: 174.24 g/mol Density: 0.975 Flash Pt: >100 °C(212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01317789 CAS Number: 121492-06-6 UN: 2735 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...