Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium tetrakis(4-fluorophenyl)borate dihydrate 98% (by HPLC)

Supplier: Alfa Aesar
Warning

H55952-06 H55952-03 H55952-MD
AAH55952-06EA 1341.06 USD InStock
AAH55952-06 AAH55952-03 AAH55952-MD
Sodium tetrakis(4-fluorophenyl)borate dihydrate 98% (by HPLC)
Sodium tetrakis(4-fluorophenyl)borate dihydrate
Sodium tetrakis(4-fluorophenyl)borate dihydrate, is used as a reagent for the two-phase titration of nonionic surfactants in the presence of anionic surfactants.

Caution: Hygroscopic. Incompatible withstrong oxidizing agents.


Formula: C₂₄H₂₀BF₄NaO₂
MW: 450,215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149598
CAS Number: 207683-22-5
Order Now
Specification Test Results
Assay (HPLC) Min. 97.5%

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty