2-Pyridyl trifluoromethanesulfonate ≥98%

Supplier: Alfa Aesar
Warning

Synonyms: Trifluoromethanesulfonic Acid 2-Pyridyl Ester2-Pyridyl Triflate

H55855-AA H55855-AC
AAH55855-AAEA 111.5 USD
AAH55855-AA AAH55855-AC
2-Pyridyl trifluoromethanesulfonate ≥98%
2-Pyridyl trifluoromethanesulfonate
Formula: C₆H₄F₃NO₃S
MW: 227.16 g/mol
Boiling Pt: 108 °C (25 mmHg)
Density: 1,477
Flash Pt: 27 °C (81 °F)
Storage Temperature: Ambient
MDL Number: MFCD00192531
CAS Number: 65007-00-3
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR