Potassium-4-acetylphenyltrifluoroborate 97%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: Potassium (4-acetylphenyl)trifluoroboranuidePotassium 4-acetylphenyltrifluoroborate

H55845-14 H55845-03 H55845-06
AAH55845-06EA 148 USD
AAH55845-06 AAH55845-14 AAH55845-03
Potassium-4-acetylphenyltrifluoroborate 97%
Potassium 4-acetylphenyltrifluoroborate
Formula: C₈H₇BF₃KO
MW: 226.05 g/mol
Melting Pt: 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD07784376
CAS Number: 252726-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR