(1,1-Dimethoxyethyl)benzene 97%

Warning

H55696-06 H55696-14
AAH55696-14EA 262.69 USD
AAH55696-14 AAH55696-06
(1,1-Dimethoxyethyl)benzene 97%
(1,1-Dimethoxyethyl)benzene
Formula: C₆H₅C(OCH₃)₂CH₃
MW: 166.22 g/mol
Boiling Pt: 95…98 °C/12 mm
Density: 1.008
Flash Pt: 67 °C(153 °F)
Storage Temperature: Ambient
MDL Number: MFCD00075617
CAS Number: 4316-35-2
EINECS: 224-334-3
https://us.vwr.com/stibo/web/std.lang.all/78/02/2297802.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR