1-(3,4-Dichlorobenzyl)piperazine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H55685-06
H55685-03
H55685-14
AAH55685-03EA
60.17
USD
AAH55685-03
AAH55685-14
AAH55685-06
1-(3,4-Dichlorobenzyl)piperazine 97%
1-(3,4-Dichlorobenzyl)piperazine
Formula:
C₁₁H₁₄Cl₂N₂ Boiling Pt: 124…128 °C/0.02 mm Density: 1.225 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03407488 CAS Number: 55513-17-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...