Pyridinium trifluoromethanesulfonate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Pyridinium Triflate
H55659-14
H55659-06
AAH55659-14EA
191.99
USD
AAH55659-14
AAH55659-06
Pyridinium trifluoromethanesulfonate 97%
Pyridinium trifluoromethanesulfonate
Formula:
C₆H₆F₃NO₃S MW: 229.17 g/mol Melting Pt: 221…223 °C Storage Temperature: Ambient |
MDL Number:
MFCD00274656 CAS Number: 52193-54-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...