4-Bromo-1-naphthaleneboronic acid 97%

Warning

Synonyms: (4-Bromonaphthalen-1-yl)boronic acid

H55625-MD H55625-03 H55625-06
AAH55625-03EA 94.15 USD
AAH55625-03 AAH55625-MD AAH55625-06
4-Bromo-1-naphthaleneboronic acid 97%
4-Bromo-1-naphthaleneboronic acid
Formula: C₁₀H₈BBrO₂
MW: 250.88 g/mol
Melting Pt: 230…260 °C
Storage Temperature: Ambient
MDL Number: MFCD08701764
CAS Number: 145965-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR