2-Mesitylenesulfonyl hydrazide 97%
|
Warning
|
Synonyms:
2,4,6-Trimethylbenzenesulfonyl Hydrazide
H55586-14
H55586-06
H55586-03
AAH55586-06EA
58.99
USD
AAH55586-06
AAH55586-14
AAH55586-03
2-Mesitylenesulfonyl hydrazide 97%
2-Mesitylenesulfonyl hydrazide
Formula:
C₉H₁₄N₂O₂S MW: 214.28 g/mol Melting Pt: 112…114 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00007585 CAS Number: 16182-15-3 EINECS: 240-319-4 UN: 1325 ADR: "4.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...