3,3'-Di-n-hexyloxacarbocyanine iodide 98%

Warning

Synonyms: 3,3'-Dihexyloxacarbocyanine iodideDiOC6(3)DiOC6(3) iodide3-Hexyl-2-[3-(3-hexyl-1,3-benzoxazol-2(3H)-ylidene)-1-propen-1-yl]-1,3-benzoxazol-3-ium iodide3-hexyl-2-[3-(3-hexyl-2(3H)benzoxazolylidene)-1-propenyl]benzoxazolium iodide

H55574-MD H55574-03 H55574-06
AAH55574-MDEA 62.92 USD
AAH55574-MD AAH55574-06 AAH55574-03
3,3'-Di-n-hexyloxacarbocyanine iodide 98%
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3)]
Formula: C₂₉H₃₇IN₂O₂
MW: 572.531 g/mol
Melting Pt: 219…221 °C
Storage Temperature: Ambient
MDL Number: MFCD00011954
CAS Number: 53213-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR