1-Methylpiperazine dihydrochloride 98%

Warning

H55536-06 H55536-14
AAH55536-06EA 23.37 USD
AAH55536-06 AAH55536-14
1-Methylpiperazine dihydrochloride 98%
1-Methylpiperazine dihydrochloride
Formula: C₅H₁₄Cl₂N₂
Melting Pt: 246…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00012755
CAS Number: 34352-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR