4-Chloro-2,5-difluorobenzoic acid 98%

Danger

H55521-14 H55521-06
AAH55521-06EA 43.33 USD
AAH55521-06 AAH55521-14
4-Chloro-2,5-difluorobenzoic acid 98%
4-Chloro-2,5-difluorobenzoic acid
Formula: C₇H₃ClF₂O₂
Melting Pt: 154…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00077480
CAS Number: 132794-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR