9,9-Di-n-octylfluorene-2,7-diboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (9,9-Dioctyl-9H-fluorene-2,7-diyl)diboronic acidBoronic acid,B,B'-(9,9-dioctyl-9H-fluorene-2,7-diyl)bis-9,9-Dioctylfluorene-2,7-diboronic acid

H55373-MD H55373-03
AAH55373-MDEA 35.18 USD
AAH55373-MD AAH55373-03
9,9-Di-n-octylfluorene-2,7-diboronic acid 97%
9,9-Di-n-octylfluorene-2,7-diboronic acid
Formula: C₂₉H₄₄B₂O₄
MW: 478.287 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03701608
CAS Number: 258865-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR