Potassium thiophene-3-trifluoroborate 97%

Warning

Synonyms: Potassium trifluoro(thiophen-3-yl)borate

H55313-03 H55313-MD H55313-06
AAH55313-03EA 65.5 USD
AAH55313-03 AAH55313-06 AAH55313-MD
Potassium thiophene-3-trifluoroborate 97%
Potassium thiophene-3-trifluoroborate
Formula: C₄H₃BF₃KS
MW: 190.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03701626
CAS Number: 192863-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR