Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Methoxy-N,N-dimethylbenzylamine ≥98%

Supplier: Alfa Aesar
Warning

H55260-06 H55260-14 H55260-03
AAH55260-06 AAH55260-14 AAH55260-03
3-Methoxy-N,N-dimethylbenzylamine ≥98%


Formula: C₁₀H₁₅NO
MW: 165,236 g/mol
Boiling Pt: 105 °C (13 mmHg)
Density: 0,984
Flash Pt: 96 °C (205 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011530
CAS Number: 15184-99-3
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty