2-Bromothiobenzamide 97%

Supplier: Thermo Scientific Chemicals
Warning

H55238-MD H55238-06 H55238-03
AAH55238-03EA 91.69 USD
AAH55238-03 AAH55238-06 AAH55238-MD
2-Bromothiobenzamide 97%
2-Bromothiobenzamide
Formula: C₇H₆BrNS
MW: 216.1 g/mol
Melting Pt: 82…86 °C
Storage Temperature: Ambient
MDL Number: MFCD04973314
CAS Number: 30216-44-5
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR