1-(2-Methylbenzyl)piperazine 97%

Supplier: Thermo Scientific Chemicals
Warning

H55203-14 H55203-03 H55203-06
AAH55203-06EA 103.77 USD
AAH55203-06 AAH55203-14 AAH55203-03
1-(2-Methylbenzyl)piperazine 97%
1-(2-Methylbenzyl)piperazine
Formula: C₁₂H₁₈N₂
Boiling Pt: 77…80 °C/0.1 mm
Density: 0.988
Storage Temperature: Ambient
MDL Number: MFCD02256026
EINECS: 226-183-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR