Tributyltin trifluoromethanesulfonate tech. 90%, Technical Grade
|
Danger
|
Synonyms:
Tributyltin triflate
H55192-MD
H55192-06
H55192-03
AAH55192-MDEA
16.73
USD
AAH55192-MD
AAH55192-06
AAH55192-03
Tributyltin trifluoromethanesulfonate tech. 90%, Technical Grade
Tributyltin trifluoromethanesulfonate
Formula:
C₁₃H₂₇F₃O₃SSn MW: 439.121 g/mol Flash Pt: 113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00043324 CAS Number: 68725-14-4 UN: 2923 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...