Di-p-tolylether 99%

Warning

Synonyms: 4,4'-Dimethyldiphenyl Ether

H55167-14 H55167-06
AAH55167-06EA 40 USD
AAH55167-06 AAH55167-14
Di-p-tolylether 99%
Di-p-tolylether
Formula: C₁₄H₁₄O
MW: 198.27 g/mol
Boiling Pt: 285 °C
Melting Pt: 47…49 °C
Flash Pt: >113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00025979
CAS Number: 1579-40-4
EINECS: 216-423-0
UN: 3077
ADR: "9",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR