DASPEI (2-(4-(Dimethylamino)styryl)-N-ethylpyridinium iodide) ≥99%

Warning

H55163-MD H55163-03
AAH55163-MDEA 120.79 USD
AAH55163-MD AAH55163-03
DASPEI (2-(4-(Dimethylamino)styryl)-N-ethylpyridinium iodide) ≥99%
DASPEI (2-(4-(Dimethylamino)styryl)-N-ethylpyridinium iodide)
Formula: C₁₇H₂₁IN₂
Melting Pt: 267 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011994
CAS Number: 3785-01-1
EINECS: 223-248-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR