1-(4-Bromobenzyl)piperazine 97%
|
Danger
|
H55145-MD
H55145-03
AAH55145-03EA
60.6
USD
AAH55145-03
AAH55145-MD
1-(4-Bromobenzyl)piperazine 97%
1-(4-Bromobenzyl)piperazine
Formula:
C₁₁H₁₅BrN₂ Boiling Pt: 112…116 °C/0.3 mm Melting Pt: 56…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD02177416 CAS Number: 91345-62-9 UN: 3259 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...