3,3'-Diethyl-9-methylthiacarbocyanine iodide 98%

Supplier: Thermo Scientific Chemicals

H55144-MD H55144-03
AAH55144-03EA 244.99 USD
AAH55144-03 AAH55144-MD
3,3'-Diethyl-9-methylthiacarbocyanine iodide 98%
3,3'-Diethyl-9-methylthiacarbocyanine iodide
Formula: C₂₂H₂₃IN₂S₂
Melting Pt: 275…285 °C
Storage Temperature: Ambient
MDL Number: MFCD00011955
CAS Number: 3065-79-0
EINECS: 221-317-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR