3-(Trimethylsilylethynyl)pyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-((Trimethylsilyl)ethynyl)pyridine
H55101-06
H55101-03
H55101-14
AAH55101-03EA
23.09
USD
AAH55101-03
AAH55101-14
AAH55101-06
3-(Trimethylsilylethynyl)pyridine 97%
3-(Trimethylsilylethynyl)pyridine
Formula:
C₁₀H₁₃NSi MW: 175.31 g/mol Boiling Pt: 229 °C Density: 0.922 Flash Pt: 94 °C(201 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01863744 CAS Number: 80673-00-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...