4-Propoxyaniline 97%

Supplier: Thermo Scientific Chemicals

Warning

H55096-03 H55096-06
AAH55096-03EA 41.1 USD
AAH55096-03 AAH55096-06
4-Propoxyaniline 97%
4-Propoxyaniline
Formula: C₉H₁₃NO
Flash Pt: 110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00043621
CAS Number: 4469-80-1
EINECS: 224-740-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR