3,5-Dimethyl-4-methoxyphenylmagnesium bromide 0.5 M in 2-MeTHF (2-Methyltetrahydrofuran)

Danger

H54781-AE
AAH54781-AEEA 319 USD
AAH54781-AE
3,5-Dimethyl-4-methoxyphenylmagnesium bromide 0.5 M in 2-MeTHF (2-Methyltetrahydrofuran)
3,5-Dimethyl-4-methoxyphenylmagnesium bromide
Formula: C₉H₁₁BrMgO
Storage Temperature: Ambient
MDL Number: MFCD13319917
CAS Number: 185416-17-5
UN: 2924
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR