3-Chlorobenzylmagnesium chloride 0.50 M in 2-MeTHF (2-Methyltetrahydrofuran)

Supplier: Thermo Scientific Chemicals
Danger

H54237-AE
AAH54237-AEEA 339.99 USD
AAH54237-AE
3-Chlorobenzylmagnesium chloride 0.50 M in 2-MeTHF (2-Methyltetrahydrofuran)
3-Chlorobenzylmagnesium chloride
Formula: C₇H₆Cl₂Mg
Storage Temperature: Ambient
MDL Number: MFCD01319894
CAS Number: 29874-01-9
UN: 2924
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR