4-Ethylphenylmagnesium bromide 0.5 M in 2-MeTHF (2-Methyltetrahydrofuran)

Danger

H54137-AE
AAH54137-AEEA 682.47 USD
AAH54137-AE
4-Ethylphenylmagnesium bromide 0.5 M in 2-MeTHF (2-Methyltetrahydrofuran)
4-Ethylphenylmagnesium bromide
Formula: C₈H₉BrMg
Storage Temperature: Ambient
MDL Number: MFCD01311467
CAS Number: 22873-28-5
UN: 2924
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR