3-bromo-4-methoxybenzenesulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Bromo-4-methoxybenzene-1-sulphonyl chloride
, 3-Bromo-4-methoxybenzene-1-sulfonyl chloride
H54015-03
H54015-MD
H54015-06
AAH54015-06EA
277.99
USD
AAH54015-06
AAH54015-MD
AAH54015-03
3-bromo-4-methoxybenzenesulfonyl chloride 97%
3-bromo-4-methoxybenzenesulfonyl chloride
Formula:
C₇H₆BrClO₃S MW: 285.536 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03989643 CAS Number: 23094-96-4 UN: 3265 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...