2-Amino-3-bromo-5-fluoropyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Bromo-5-fluoro-2-pyridylamine 3-Bromo-5-fluoropyridin-2-amine

H37257-03 H37257-06
AAH37257-06EA 262.99 USD
AAH37257-06 AAH37257-03
2-Amino-3-bromo-5-fluoropyridine 97%
2-Amino-3-bromo-5-fluoropyridine
Formula: C₅H₄BrFN₂
Boiling Pt: 63…65 °C
Storage Temperature: Ambient
MDL Number: MFCD03092924
CAS Number: 869557-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR