4-Methoxy-3-methyl-5-(trifluoromethyl)benzamide 97%

Warning

Synonyms: Benzamide, 4-methoxy-3-methyl-5-(trifluoromethyl)-

H35873-06 H35873-03
AAH35873-06EA 571.65 USD
AAH35873-06 AAH35873-03
4-Methoxy-3-methyl-5-(trifluoromethyl)benzamide 97%
4-Methoxy-3-methyl-5-(trifluoromethyl)benzamide
Formula: C₁₀H₁₀F₃NO₂
MW: 233.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22682823
CAS Number: 1431329-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR