5-Methyl-2-(trifluoromethoxy)aniline 97%

Warning

Synonyms: 5-Methyl-2-(trifluoromethoxy)benzenamine6-(Trifluoromethoxy)-m-toluidineBenzenamine, 5-methyl-2-(trifluoromethoxy)-

H35752-MD
AAH35752-MDEA 183.15 USD
AAH35752-MD
5-Methyl-2-(trifluoromethoxy)aniline 97%
5-Methyl-2-(trifluoromethoxy)aniline
Formula: C₈H₈F₃NO
MW: 191.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18399696
CAS Number: 151276-15-2
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR