[(S,S)-Teth-MsDPEN-RuCl] (18,5% Ru)
|
Warning
|
Synonyms:
Chloro{(1S,2S)-1,2-diphenyl-1-[(3-(?6-phenyl)propyl)amino]-2-(methylsulfonylamido)}ruthenium(II) RuCl[(S,S)2teth-MsDpen]
H35726-03
AAH35726-03EA
493.95
USD
AAH35726-03
[(S,S)-Teth-MsDPEN-RuCl] (18,5% Ru)
[(S,S)-Teth-MsDPEN-RuCl]
Formula:
C₂₄H₂₇ClN₂O₂RuS MW: 544.072 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22988933 CAS Number: 1437326-26-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...