4-Aminopyrimidine-5-carboxaldehyde 97%

Warning

Synonyms: 4-Aminopyrimidine-5-carbaldehyde

H35706-03 H35706-MD
AAH35706-MDEA 174.27 USD
AAH35706-MD AAH35706-03
4-Aminopyrimidine-5-carboxaldehyde 97%
4-Aminopyrimidine-5-carboxaldehyde
Formula: C₅H₅N₃O
MW: 123.115 g/mol
Melting Pt: 192…200 °C
Storage Temperature: Ambient
MDL Number: MFCD09991893
CAS Number: 16357-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR