DPPF RuCl2 AMPY

Warning

H35605-03
AAH35605-03EA 130.98 USD
AAH35605-03
DPPF RuCl2 AMPY
DPPF RuCl2 AMPY
Formula: C₄₀H₃₆Cl₂FeN₂P₂Ru
MW: 834.505 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22988930
CAS Number: 1287255-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR