4-(Pentafluorothio)benzonitrile 97%

Warning

H35537-06 H35537-03
AAH35537-06EA 295.26 USD
AAH35537-06 AAH35537-03
4-(Pentafluorothio)benzonitrile 97%
4-(Pentafluorothio)benzonitrile
Formula: C₇H₄F₅NS
Storage Temperature: Ambient
MDL Number: MFCD03788521
CAS Number: 401892-85-1
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR