2-Chloro-5-(pentafluorothio)benzaldehyde 97%

Warning

H35247-03 H35247-MD
AAH35247-03EA 183.15 USD
AAH35247-03 AAH35247-MD
2-Chloro-5-(pentafluorothio)benzaldehyde 97%
2-Chloro-5-(pentafluorothio)benzaldehyde
Formula: C₇H₄ClF₅OS
MW: 266.61
Storage Temperature: Ambient
MDL Number: MFCD22988924
CAS Number: 1431329-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR