Bis(diphenylphosphinomethyl)phenylphosphine oxide 95%
|
Warning
|
Synonyms:
1,1,3,5,5-Pentaphenyl-1,3,5-triphosphapentane trioxide
H35194-06
H35194-MD
H35194-03
AAH35194-06EA
563.88
USD
AAH35194-06
AAH35194-MD
AAH35194-03
Bis(diphenylphosphinomethyl)phenylphosphine oxide 95%
Bis(diphenylphosphinomethyl)phenylphosphine oxide
Formula:
C₃₂H₂₉OP₃ MW: 522.504 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00482218 CAS Number: 21851-89-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...