3-(4-Chloro-3-ethoxy-2-fluorophenyl)propionic acid 97%

Warning

Synonyms: 3-(4-Chloro-3-ethoxy-2-fluorophenyl)propanoic acid4-Chloro-3-ethoxy-2-fluorobenzenepropanoic acidBenzenepropanoic acid, 4-chloro-3-ethoxy-2-fluoro-

H34353-03 H34353-MD
AAH34353-03EA 198.69 USD
AAH34353-03 AAH34353-MD
3-(4-Chloro-3-ethoxy-2-fluorophenyl)propionic acid 97%
3-(4-Chloro-3-ethoxy-2-fluorophenyl)propionic acid
Formula: C₁₁H₁₂ClFO₃
MW: 246.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687220
CAS Number: 1323966-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR