(6-Nitro-2-benzothiazolyl)thiourea ≥97%
|
Warning
|
Synonyms:
1-(6-Nitro-1,3-benzothiazol-2-yl)thiourea
H34335-03
H34335-06
H34335-MD
AAH34335-03EA
159.51
USD
AAH34335-03
AAH34335-06
AAH34335-MD
(6-Nitro-2-benzothiazolyl)thiourea ≥97%
(6-Nitro-2-benzothiazolyl)thiourea
Formula:
C₈H₆N₄O₂S₂ MW: 254.29 g/mol Melting Pt: 245…249 °C Storage Temperature: Ambient |
MDL Number:
MFCD22682811 CAS Number: 56159-94-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...