4-Fluoro-2-(trifluoromethoxy)benzonitrile 97%

Warning

H34260-06 H34260-03
AAH34260-03EA 123.21 USD
AAH34260-03 AAH34260-06
4-Fluoro-2-(trifluoromethoxy)benzonitrile 97%
4-Fluoro-2-(trifluoromethoxy)benzonitrile
Formula: C₈H₃F₄NO
Storage Temperature: Ambient
MDL Number: MFCD19687191
CAS Number: 1323966-32-0
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR