3-Chloro-5-(trifluoromethyl)picolinic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Chloro-5-(trifluoromethyl)picolinic acid
, 3-Chloro-5-(trifluoromethyl)-2-pyridinecarboxylic acid
, 3-Chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid
H34258-03
H34258-MD
AAH34258-MDEA
86.02
USD
AAH34258-MD
AAH34258-03
3-Chloro-5-(trifluoromethyl)picolinic acid 97%
3-Chloro-5-(trifluoromethyl)picolinic acid
Formula:
C₇H₃ClF₃NO₂ MW: 225.60 g/mol Melting Pt: 141…143 °C Storage Temperature: Ambient |
MDL Number:
MFCD00277482 CAS Number: 80194-68-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...