3-[4-Fluoro-3-methyl-5-(trifluoromethyl)phenyl]propionic acid 97%

Warning

Synonyms: 3-(4-Fluoro-3-methyl-5-(trifluoromethyl)phenyl)propanoic acid

H34163-MD H34163-03
AAH34163-MDEA 46.2 USD
AAH34163-MD AAH34163-03
3-[4-Fluoro-3-methyl-5-(trifluoromethyl)phenyl]propionic acid 97%
3-[4-Fluoro-3-methyl-5-(trifluoromethyl)phenyl]propionic acid
Formula: C₁₁H₁₀F₄O₂
MW: 250.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201076
CAS Number: 1373920-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR