2-Methoxy-5-(pentafluorothio)benzylamine 97%

Danger

Synonyms: 2-Methoxy-5-(pentafluorosulfur)benzylamine

H34149-03 H34149-MD
AAH34149-03EA 247.53 USD
AAH34149-03 AAH34149-MD
2-Methoxy-5-(pentafluorothio)benzylamine 97%
2-Methoxy-5-(pentafluorothio)benzylamine
Formula: C₈H₁₀F₅NOS
MW: 263.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652492
CAS Number: 1211516-76-5
UN: 3259
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR