Ethyl-4-bromo-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

H34065-03 H34065-MD
AAH34065-03EA 428.99 USD
AAH34065-03 AAH34065-MD
Ethyl-4-bromo-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate 97%
Ethyl-4-bromo-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate
Formula: C₇H₆BrF₃N₂O₂
MW: 287.036 g/mol
Melting Pt: 131…133 °C
Storage Temperature: Ambient
MDL Number: MFCD13193102
CAS Number: 1240278-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR