6-Acetylpicolinic acid 95%

Warning

Synonyms: 6-Acetylpyridine-2-carboxylic acid

H34059-MD
AAH34059-MDEA 139.86 USD
AAH34059-MD
6-Acetylpicolinic acid 95%
6-Acetylpicolinic acid
Formula: C₈H₇NO₃
Melting Pt: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD09800364
CAS Number: 122637-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR