2,4-Dichloro-3-formylpyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,4-Dichloropyridine-3-carboxaldehyde
, 2,4-Dichloro-3-pyridinecarboxaldehyde
, 2,4-Dichloronicotinaldehyde
, 2,4-Dichloro-3-formylpyridine
H34053-MD
H34053-03
AAH34053-03EA
154.99
USD
AAH34053-03
AAH34053-MD
2,4-Dichloro-3-formylpyridine 97%
2,4-Dichloro-3-formylpyridine
Formula:
C₆H₃Cl₂NO MW: 176.00 g/mol Melting Pt: 72…75 °C Storage Temperature: Ambient |
MDL Number:
MFCD07437909 CAS Number: 134031-24-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...