2,6-Dichloro-3-pyridinemethanol 95%

Warning

Synonyms: (2,6-Dichloropyridin-3-yl)methanol

H33971-03 H33971-MD
AAH33971-03EA 143.06 USD
AAH33971-03 AAH33971-MD
2,6-Dichloro-3-pyridinemethanol 95%
2,6-Dichloro-3-pyridinemethanol
Formula: C₆H₅Cl₂NO
MW: 178.012 g/mol
Melting Pt: 67…69 °C
Storage Temperature: Ambient
MDL Number: MFCD11036367
CAS Number: 55304-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR